Assay Details
Gene Symbol: Hus1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153072.2 | NP_694712.1 | 1 | 826 | 110 | |
GenBank mRNA | AF508546.1 | - | 1 | 739 | 110 | |
AK133107.1 | - | 1 | 828 | 110 | ||
BC125607.1 | - | 1 | 819 | 40130122 | 110 | |
BC125609.1 | - | 1 | 819 | 40130124 | 110 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P258359 | AF508546.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P258359 | AK133107.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P258359 | BC125607.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P258359 | NM_153072.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P258359 | BC125609.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P258359 | AF508546.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P258359 | AK133107.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P258359 | BC125607.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P258359 | NM_153072.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P258359 | BC125609.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P107322 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P258359 | AF508546.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P258359 | AK133107.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P258359 | BC125607.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P258359 | NM_153072.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P258359 | BC125609.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P258359 | AF508546.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P258359 | AK133107.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P258359 | BC125607.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P258359 | NM_153072.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P258359 | BC125609.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P258359 | AF508546.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P258359 | AK133107.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P258359 | BC125607.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P258359 | NM_153072.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P258359 | BC125609.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P258359 | AF508546.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P258359 | AK133107.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P258359 | BC125607.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P258359 | NM_153072.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P258359 | BC125609.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135365 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28920191 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28920146 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28920189 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28920194 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28920148 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28920178 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28920192 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28920154 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28920140 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28920174 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28920153 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28920176 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28920183 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28920139 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28920164 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28920198 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28920166 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28920200 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28920160 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28920143 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28920142 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28920144 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28920167 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28920150 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28920138 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28920180 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28920197 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28920170 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28920200 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28920160 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10760314 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10763565 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28920155 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28920162 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10512963 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28920186 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28920138 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28920185 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28920157 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28920194 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28920172 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28920173 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28920174 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28920170 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28920139 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10763566 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28920198 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28920178 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28920149 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28920182 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28920146 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28920183 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28920177 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28920191 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28920148 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28920156 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28920187 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10760324 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10794858 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28920164 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11026494 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28920180 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28920143 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28920192 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28920150 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28920197 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28920189 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10856095 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10408520 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408521 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408522 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10408519 | - | - |
Affymetrix Mouse Clariom S Array | PSR1300010815.mm.1 | NM_153072.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300010815.mm.1 | NM_153072.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300010815.mm.1 | TC1300001739.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 808929 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 2900592 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002900592 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1240847 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1240847 | - | - |
mouse-6 v1.1 (Illumina) | scl44138.1.1_100-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2900592 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl44138.1.1_100-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2900592 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1240847 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1240847 | - | - |
