Assay Details
| Gene Symbol: Egfl8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_152922.3 | NP_690886.3 | 7 - 8 | 774 | 66 | |
| GenBank mRNA | BC139484.1 | - | 7 - 8 | 743 | 8862506 | 66 |
| BC141312.1 | - | 7 - 8 | 743 | 9055924 | 66 | |
| BC145632.1 | - | 7 - 8 | 743 | 9054200 | 66 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Egfl8
Genomic Map