Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700006573.mm.1 |
NM_001110101.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006573.mm.1 |
NM_001110101.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006573.mm.1 |
NM_001204273.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006573.mm.1 |
NM_001204273.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006573.mm.1 |
NM_030597.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006573.mm.1 |
NM_030597.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006573.mm.1 |
TC1700000657.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700006576.mm.1 |
NM_001110101.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006576.mm.1 |
NM_001110101.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006576.mm.1 |
NM_001204273.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006576.mm.1 |
NM_001204273.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006576.mm.1 |
NM_001204274.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006576.mm.1 |
NM_001204274.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006576.mm.1 |
NM_030597.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006576.mm.1 |
NM_030597.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006576.mm.1 |
TC1700000657.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700006575.mm.1 |
NM_001110101.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006575.mm.1 |
NM_001110101.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006575.mm.1 |
NM_001204273.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006575.mm.1 |
NM_001204273.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006575.mm.1 |
NM_001204274.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006575.mm.1 |
NM_001204274.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006575.mm.1 |
NM_030597.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006575.mm.1 |
NM_030597.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006575.mm.1 |
TC1700000657.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700006577.mm.1 |
NM_001110101.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006577.mm.1 |
NM_001110101.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006577.mm.1 |
NM_001204273.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006577.mm.1 |
NM_001204273.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006577.mm.1 |
NM_001204274.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006577.mm.1 |
NM_001204274.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006577.mm.1 |
NM_030597.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006577.mm.1 |
NM_030597.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006577.mm.1 |
TC1700000657.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700006574.mm.1 |
NM_001110101.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006574.mm.1 |
NM_001110101.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006574.mm.1 |
NM_001204273.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006574.mm.1 |
NM_001204273.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006574.mm.1 |
NM_001204274.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006574.mm.1 |
NM_001204274.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006574.mm.1 |
NM_030597.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006574.mm.1 |
NM_030597.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006574.mm.1 |
TC1700000657.mm.1 |
MTA TC |