Assay Details
| Gene Symbol: Smkr-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040263.1 | 1 - 2 | 134 | 83 | ||
| GenBank mRNA | AK006282.1 | - | 1 - 2 | 112 | 83 | |
| BY706362.1 | - | 1 - 2 | 106 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P142109 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P499948 | AK076883.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P499948 | AV205010.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P499948 | AK006282.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P499948 | AK076883.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P499948 | AV205010.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P499948 | AK006282.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P142109 | - | - |
| Whole Mouse Genome G4122A | A_52_P499948 | AK076883.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P499948 | AV205010.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P499948 | AK006282.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P499948 | AK076883.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P499948 | AV205010.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P499948 | AK006282.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120961 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P142109 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05723211 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24329515 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24329520 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24329548 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24329525 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05138180 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05348300 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24329621 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24329524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24329536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24329512 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05809962 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24329535 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24329618 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05175691 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05258147 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24329517 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24329555 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24329545 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24329550 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24329513 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24329533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24329523 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24329620 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24329553 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24329495 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24329529 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24329541 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24329516 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05052628 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05537862 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24329521 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24329501 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05052625 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24329622 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05339134 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24329506 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05655341 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05703875 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24329551 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24329554 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05348306 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05348323 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24329540 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24329623 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05053575 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24329558 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05877530 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05646757 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24329531 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24329544 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1438280_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1438281_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1438281_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1438280_at | - | - |
| Mouse Genome 430 2.0 Array | 1438281_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1438280_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76419_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76419_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600001796.mm.1 | TC0600000255.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600001796.mm.1 | NR_040263.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600001796.mm.1 | NR_040263.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600001796.mm.1 | NR_040264.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600001796.mm.1 | NR_040264.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600001798.mm.1 | TC0600000255.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600001798.mm.1 | NR_040263.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600001798.mm.1 | NR_040263.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600001798.mm.1 | NR_040264.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600001798.mm.1 | NR_040264.1 | Refseq |
| Mouse Expression Set 430 | 1438281_x_at | - | - |
| Mouse Expression Set 430 | 1438281_PM_x_at | - | - |
| Mouse Expression Set 430 | 1438280_at | - | - |
| Mouse Expression Set 430 | 1438280_PM_at | - | - |
| Murine Genome U74v2 Set | 165828_r_at | - | - |
| Murine Genome U74v2 Set | 167943_PM_f_at | - | - |
| Murine Genome U74v2 Set | 167943_f_at | - | - |
| Murine Genome U74v2 Set | 165828_PM_r_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438281_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438281_x_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438280_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1438280_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 100630504 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2677217 | - | - |

Smkr-ps
Genomic Map