Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00020292.mm.1 |
NM_001093750.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020292.mm.1 |
NM_001093750.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020292.mm.1 |
TC0X00003241.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00020290.mm.1 |
NM_001093750.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020290.mm.1 |
NM_001093750.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020290.mm.1 |
TC0X00003241.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00020291.mm.1 |
NM_001093750.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020291.mm.1 |
NM_001093750.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020291.mm.1 |
TC0X00003241.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00020287.mm.1 |
NM_001093750.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020287.mm.1 |
NM_001093750.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020287.mm.1 |
TC0X00003241.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00020288.mm.1 |
NM_001093750.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020288.mm.1 |
NM_001093750.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00020288.mm.1 |
TC0X00003241.mm.1 |
MTA TC |