Assay Details
| Gene Symbol: 4933400A11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015979.1 | - | 1 - 2 | 77 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK015979.1 transcripts.
Set Membership: |
|

4933400A11Rik
Genomic Map