Assay Details
| Gene Symbol: Irak3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028679.3 | NP_082955.2 | 5 - 6 | 702 | 74 | |
| GenBank mRNA | AF461763.1 | - | 5 - 6 | 682 | 74 | |
| AJ440757.2 | - | 5 - 6 | 624 | 74 | ||
| AK029057.1 | - | 5 - 6 | 703 | 74 | ||
| AK049210.1 | - | 5 - 6 | 733 | 74 | ||
| BC120829.1 | - | 5 - 6 | 663 | 40129752 | 74 | |
| BC120831.1 | - | 5 - 6 | 663 | 40129754 | 74 | |
| BC144835.1 | - | 5 - 6 | 663 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Irak3
Genomic Map