Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300009693.mm.1 |
NM_181754.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300009693.mm.1 |
NM_181754.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300009693.mm.1 |
TC1300001526.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300009694.mm.1 |
NM_181754.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300009694.mm.1 |
NM_181754.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300009694.mm.1 |
TC1300001526.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300009695.mm.1 |
NM_181754.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300009695.mm.1 |
NM_181754.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300009695.mm.1 |
TC1300001526.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300009696.mm.1 |
NM_181754.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300009696.mm.1 |
NM_181754.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300009696.mm.1 |
TC1300001526.mm.1 |
MTA TC |