Assay Details
Gene Symbol: 9430018G01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045988.1 | 1 - 2 | 341 | 91 | ||
GenBank mRNA | AK034646.1 | - | 1 - 2 | 341 | 91 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | thiamine pyrophosphokinase | Tpk1 | Anti-sense | Chr. 6 - 43344999 - 43666484 on NCBI Build GRCm38 | ||
Downstream | contactin associated protein-like 2 | Cntnap2 |
5430425M22Rik, Caspr2, mKIAA0868 |
1.6mb | Sense | Chr. 6 - 45060061 - 47301388 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
