Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600003714.mm.1 |
TC0600000460.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003714.mm.1 |
NR_045169.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003714.mm.1 |
NR_045169.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003717.mm.1 |
TC0600000460.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003717.mm.1 |
NR_045169.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003717.mm.1 |
NR_045169.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003715.mm.1 |
TC0600000460.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003715.mm.1 |
NR_045169.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003715.mm.1 |
NR_045169.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003719.mm.1 |
TC0600000460.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600003719.mm.1 |
NR_045169.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600003719.mm.1 |
NR_045169.1 |
Refseq |
Murine Genome U74v2 Set |
169727_i_at |
- |
- |
Murine Genome U74v2 Set |
169727_PM_i_at |
- |
- |