Assay Details
Gene Symbol: Btn2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289615.1 | NP_001276544.1 | 6 - 7 | 1117 | 56 | |
NM_175938.3 | NP_787952.2 | 6 - 7 | 1176 | 56 | ||
GenBank mRNA | AK051061.1 | - | 6 - 7 | 1163 | 56 | |
AY177686.1 | - | 6 - 7 | 1159 | 56 | ||
BC139760.1 | - | 6 - 7 | 1003 | 40058494 | 56 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
