Assay Details
| Gene Symbol: Gm11377 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK085428.1 | - | 117 | 71 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | forkhead box Q1 | Foxq1 |
HFH-1, Hfh1, Hfh1l, sa |
-3.8kb | Sense | Chr. 13 - 31558170 - 31560976 on NCBI Build GRCm38 |
| Downstream | forkhead box F2 | Foxf2 |
FREAC2, Fkh20, LUN |
59.8kb | Sense | Chr. 13 - 31625816 - 31631406 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm11377
Genomic Map