Assay Details
Gene Symbol: Prl7b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029355.2 | NP_083631.1 | 4 - 5 | 417 | 81 | |
GenBank mRNA | AF525156.1 | - | 4 - 5 | 417 | 81 | |
AK005458.1 | - | 4 - 5 | 418 | 81 | ||
BC116772.1 | - | 4 - 5 | 394 | 40126091 | 81 | |
BC119183.1 | - | 4 - 5 | 394 | 8733932 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2164258 | AK005458.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2164258 | AF525156.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2164258 | BC116772.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2164258 | BC119183.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2164258 | NM_029355.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 867311 | AK050104.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 867311 | AK086578.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 371204 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 10802 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 330990 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 937209 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 25236 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 118493 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 117506 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P121679 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2164258 | AK005458.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2164258 | AF525156.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2164258 | BC116772.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2164258 | BC119183.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2164258 | NM_029355.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902856 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902857 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902870 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902862 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902865 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902873 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902882 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902859 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902879 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902861 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902853 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902854 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902850 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902855 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902867 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902866 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902876 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902872 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902847 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902848 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902875 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902871 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902881 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902852 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902851 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902878 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28902849 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P121679 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07818921 | - | - |
Whole Mouse Genome G4122A | A_51_P458338 | AK005458.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P458338 | AF525156.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P458338 | BC116772.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P458338 | BC119183.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P458338 | NM_029355.2 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P121679 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10968690 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10968691 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10968682 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10968689 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10968685 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10968679 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10968687 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902808 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902802 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902799 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902883 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902779 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902865 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902886 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902871 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902887 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902810 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902862 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902824 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902819 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902888 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902848 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902856 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902881 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902875 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902793 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902867 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902841 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902873 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902812 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902823 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902827 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902813 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902790 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902845 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902844 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902797 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902825 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902821 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902815 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902781 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902782 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902818 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902870 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902831 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902853 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902773 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902872 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902837 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902878 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902805 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902834 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902857 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902786 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902829 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902879 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902801 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902839 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902807 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902843 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902842 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902800 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902804 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902816 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28902851 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902867 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902823 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902865 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10968679 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902855 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10525032 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902862 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902881 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902882 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902834 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902841 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902808 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902870 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902816 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902837 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902878 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902818 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902872 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902856 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902842 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902866 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902873 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902812 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902805 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902845 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10878517 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902813 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902839 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902853 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902848 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902838 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902807 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902854 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902879 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902871 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902861 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902859 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902831 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10878513 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902857 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902852 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902815 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902847 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902829 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10878514 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902804 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902824 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902827 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902810 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902851 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902843 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28902875 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10408413 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408414 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408417 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408418 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408415 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408419 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408416 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1429826_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10408412 | - | - |
Mouse Genome 430 2.0 Array | 1429826_at | - | - |
Mouse Genome 430 2.0 Array | 1429826_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75596_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75596_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1300010538.mm.1 | NM_029355.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300010538.mm.1 | NM_029355.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300010538.mm.1 | TC1300001694.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300010537.mm.1 | NM_029355.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300010537.mm.1 | NM_029355.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300010537.mm.1 | TC1300001694.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300010539.mm.1 | NM_029355.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300010539.mm.1 | NM_029355.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300010539.mm.1 | TC1300001694.mm.1 | MTA TC |
Mouse Expression Set 430 | 1429826_PM_at | - | - |
Mouse Expression Set 430 | 1429826_at | - | - |
Murine Genome U74v2 Set | 165880_f_at | - | - |
Murine Genome U74v2 Set | 170142_at | - | - |
Murine Genome U74v2 Set | 165880_PM_f_at | - | - |
Murine Genome U74v2 Set | 170142_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1429826_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1429826_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 464769 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2748492 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2748492 | - | - |
