Assay Details
Gene Symbol: Gm15245 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK079668.1 | - | 4 - 5 | 429 | 72 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | holocytochrome c synthetase | Hccs | -152.0kb | Anti-sense | Chr. X - 169311530 - 169320343 on NCBI Build GRCm38 | |
Downstream | midline 1 | Mid1 |
61B3-R, DXHXS1141, Fxy, Trim18 |
212.2kb | Sense | Chr. Y - 90718657 - 90765072 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
