Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1800008012.mm.1 |
NM_001146292.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800008012.mm.1 |
NM_001146292.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800008012.mm.1 |
NM_001146293.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800008012.mm.1 |
NM_001146293.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800008012.mm.1 |
NM_001146294.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800008012.mm.1 |
NM_001146294.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800008012.mm.1 |
NM_001174074.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800008012.mm.1 |
NM_001174074.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800008012.mm.1 |
NM_133195.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800008012.mm.1 |
NM_133195.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1800008012.mm.1 |
TC1800001088.mm.1 |
MTA TC |