Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500002404.mm.1 |
NM_175009.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002404.mm.1 |
NM_175009.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002404.mm.1 |
TC1500000351.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500002405.mm.1 |
NM_175009.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002405.mm.1 |
NM_175009.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002405.mm.1 |
TC1500000351.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500002403.mm.1 |
NM_175009.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002403.mm.1 |
NM_175009.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002403.mm.1 |
TC1500000351.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500002406.mm.1 |
NM_175009.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002406.mm.1 |
NM_175009.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002406.mm.1 |
TC1500000351.mm.1 |
MTA TC |