Assay Details
| Gene Symbol: Gls2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033264.3 | NP_001028436.2 | 10 - 11 | 1091 | 69 | |
| NM_001285777.1 | NP_001272706.1 | 9 - 10 | 1011 | 69 | ||
| NM_001285779.1 | NP_001272708.1 | 10 - 11 | 1120 | 69 | ||
| GenBank mRNA | AK039618.1 | - | 9 - 10 | 899 | 69 | |
| AK165896.1 | - | 10 - 11 | 1037 | 69 | ||
| AK168761.1 | - | 3 - 4 | 223 | 69 | ||
| AK220231.1 | - | 10 - 11 | 1043 | 69 | ||
| BC141115.1 | - | 10 - 11 | 1091 | 9055727 | 69 | |
| EU770627.1 | - | 10 - 11 | 1029 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gls2
Genomic Map