Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500001657.mm.1 |
NM_018755.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001657.mm.1 |
NM_018755.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001657.mm.1 |
NM_176073.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001657.mm.1 |
NM_176073.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001657.mm.1 |
TC1500000222.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500001655.mm.1 |
NM_018755.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001655.mm.1 |
NM_018755.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001655.mm.1 |
NM_176073.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001655.mm.1 |
NM_176073.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001655.mm.1 |
TC1500000222.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500001656.mm.1 |
NM_018755.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001656.mm.1 |
NM_018755.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001656.mm.1 |
NM_176073.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001656.mm.1 |
NM_176073.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001656.mm.1 |
TC1500000222.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500001658.mm.1 |
NM_018755.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001658.mm.1 |
NM_018755.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001658.mm.1 |
NM_176073.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001658.mm.1 |
NM_176073.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500001658.mm.1 |
TC1500000222.mm.1 |
MTA TC |