Assay Details
| Gene Symbol: Gnai1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010305.1 | NP_034435.1 | 3 - 4 | 551 | 67 | |
| GenBank mRNA | BC032995.1 | - | 3 - 4 | 493 | 67 | |
| BC138862.1 | - | 3 - 4 | 431 | 8861884 | 67 | |
| BC138865.1 | - | 3 - 4 | 431 | 8861887 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gnai1
Genomic Map