Assay Details
| Gene Symbol: Hyal4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014599.1 | - | 42 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK014599.1 transcripts.
Set Membership: |
|

Hyal4
Genomic Map