Assay Details
| Gene Symbol: Hyal4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029848.1 | NP_084124.1 | 3 - 4 | 1162 | 74 | |
| GenBank mRNA | AB524072.1 | - | 2 - 3 | 1025 | 74 | |
| BC125402.1 | - | 3 - 4 | 1162 | 40129917 | 74 | |
| BC132096.1 | - | 3 - 4 | 1162 | 40130373 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2052340 | BC132096.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2052340 | AK014599.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2052340 | BC125402.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2052340 | NM_029848.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 903953 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 648284 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 113407 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 852635 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 52157 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 587159 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 229324 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 11345 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 69668 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 312738 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 45342 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 108123 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136920 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P179330 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2052340 | BC132096.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2052340 | AK014599.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2052340 | BC125402.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2052340 | NM_029848.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136920 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05837558 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P179330 | - | - |
| Whole Mouse Genome G4122A | A_51_P448114 | AK014599.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136920 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P179330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303401 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303514 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303394 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303355 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303499 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303405 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303481 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303346 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303467 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24303372 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303356 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05565035 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303362 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303335 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303355 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303358 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303369 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303346 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303367 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303370 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303372 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05165968 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303337 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303350 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303398 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303403 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303340 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05153764 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05089675 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303387 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05309483 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05511074 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303332 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303352 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303396 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05089673 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303384 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303391 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303343 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303361 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303386 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303364 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05239560 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303399 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303338 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303354 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303376 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05309485 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303365 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303357 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303374 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303380 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303381 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303405 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P24303348 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10536726 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10536724 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10536725 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10536727 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1429990_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10536722 | - | - |
| Mouse Genome 430 2.0 Array | 1429990_at | - | - |
| Mouse Genome 430 2.0 Array | 1429990_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0600001297.mm.1 | TC0600000204.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0600001297.mm.1 | NM_029848.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0600001297.mm.1 | NM_029848.1 | Refseq |
| Mouse Expression Set 430 | 1429990_at | - | - |
| Mouse Expression Set 430 | 1429990_PM_at | - | - |
| Murine Genome U74v2 Set | 167517_PM_r_at | - | - |
| Murine Genome U74v2 Set | 167517_r_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429990_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1429990_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_2782392 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2782392 | - | - |

Hyal4
Genomic Map