Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700015224.mm.1 |
TC0700001791.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700015224.mm.1 |
NM_029842.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015224.mm.1 |
NM_029842.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015225.mm.1 |
TC0700001791.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700015225.mm.1 |
NM_029842.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015225.mm.1 |
NM_029842.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015226.mm.1 |
TC0700001791.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700015226.mm.1 |
NM_029842.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700015226.mm.1 |
NM_029842.5 |
Refseq |