Assay Details
Gene Symbol: Lrrc27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143755.1 | NP_001137227.1 | 2 - 3 | 235 | 101 | |
GenBank mRNA | AI614174.1 | - | 2 - 3 | 235 | 101 | |
AK006931.1 | - | 2 - 3 | 240 | 101 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001143755.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
