Assay Details
| Gene Symbol: Dchs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162943.1 | NP_001156415.1 | 3 - 4 | 2272 | 64 | |
| GenBank mRNA | AK048080.1 | - | 3 - 4 | 2275 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dchs1
Genomic Map