Assay Details
| Gene Symbol: Gpx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010343.2 | NP_034473.2 | 3 - 4 | 386 | 114 | |
| GenBank mRNA | AK029731.1 | - | 3 - 4 | 387 | 114 | |
| BC100749.1 | - | 3 - 4 | 386 | 40046457 | 114 | |
| BC103588.1 | - | 3 - 4 | 386 | 40045838 | 114 | |
| BC103589.1 | - | 3 - 4 | 386 | 40045837 | 114 | |
| BC105648.1 | - | 3 - 4 | 386 | 40046458 | 114 | |
| X53780.1 | - | 2 - 3 | 222 | 114 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gpx5
Genomic Map