Assay Details
| Gene Symbol: Zscan12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016684.2 | NP_057893.2 | 1 - 2 | 55 | 150 | |
| GenBank mRNA | AK075886.1 | - | 1 - 2 | 49 | 150 | |
| AK129138.1 | - | 1 - 2 | 55 | 150 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK075886.1, NM_016684.2 transcripts.
Set Membership: |
|

Zscan12
Genomic Map