Assay Details
Gene Symbol: Trim68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK089546.1 | - | 1 - 2 | 191 | 65 | |
AK155267.1 | - | 1 - 2 | 191 | 65 | ||
AK164544.1 | - | 1 - 2 | 136 | 65 | ||
BC034249.1 | - | 1 - 2 | 119 | 3675490 | 65 | |
BY014556.1 | - | 1 - 2 | 157 | 65 | ||
BY018445.1 | - | 1 - 2 | 194 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC034249.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
