Assay Details
Gene Symbol: Trim68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK089546.1 | - | 3 - 4 | 749 | 59 | |
AK155267.1 | - | 3 - 4 | 749 | 59 | ||
AK164544.1 | - | 3 - 4 | 694 | 59 | ||
BC034249.1 | - | 3 - 4 | 677 | 3675490 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P490678 | BC034249.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P490678 | AK155267.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P490678 | AK164544.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P490678 | BB818918.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P490678 | BC026718.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P490678 | BC034249.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P490678 | AK155267.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P490678 | AK164544.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P490678 | BB818918.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P490678 | BC026718.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 835618 | AK046648.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 835618 | BC052061.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 835618 | U34361.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 835618 | XM_001477293.1 | refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 835618 | XM_001478017.1 | refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 823412 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 108440 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 940753 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 636665 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 891713 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 75682 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 79992 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P111119 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P490678 | BC034249.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P490678 | AK155267.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P490678 | AK164544.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P490678 | BB818918.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P490678 | BC026718.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P490678 | BC034249.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P490678 | AK155267.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P490678 | AK164544.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P490678 | BB818918.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P490678 | BC026718.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P490678 | BC034249.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P490678 | AK155267.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P490678 | AK164544.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P490678 | BB818918.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P490678 | BC026718.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P490678 | BC034249.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P490678 | AK155267.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P490678 | AK164544.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P490678 | BB818918.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P490678 | BC026718.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380110 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380118 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380101 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380108 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380104 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380117 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380150 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380123 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380115 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380127 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380121 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380099 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380122 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380130 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380129 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380103 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380124 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380112 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380126 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380107 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25380120 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P111119 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06305525 | - | - |
Whole Mouse Genome G4122A | A_51_P490678 | BC034249.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P490678 | AK155267.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P490678 | AK164544.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P490678 | BB818918.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P490678 | BC026718.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P490678 | BC034249.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P490678 | AK155267.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P490678 | AK164544.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P490678 | BB818918.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P490678 | BC026718.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112787 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127323 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P111119 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06262918 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06262926 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06262919 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06262934 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06262927 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06262932 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06262935 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380107 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380105 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380077 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380027 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380120 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380053 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380029 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380099 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380104 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380080 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380064 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379989 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380047 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380062 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379996 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380124 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380138 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380148 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380143 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380055 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380117 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380123 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380122 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380108 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380032 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380075 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380110 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380140 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380073 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380006 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380016 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380121 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379994 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380083 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380035 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380076 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380132 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380092 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380135 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380011 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380037 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379987 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379992 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380044 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380112 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380005 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380015 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380051 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380017 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380127 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380095 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380101 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380024 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380071 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380049 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380059 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380003 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380146 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380086 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380013 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380126 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380007 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380039 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380043 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380034 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380048 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380046 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380115 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380089 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380068 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380004 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379999 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380001 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380091 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379993 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380103 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380133 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380009 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380066 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380096 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380142 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380026 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25379991 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380129 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380082 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380023 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380141 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380118 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380041 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25380020 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380103 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380073 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380095 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380115 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380096 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380089 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380083 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380032 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06502435 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380041 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380105 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06468857 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06454860 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05951961 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380127 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380037 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06454871 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380094 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380098 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380077 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380128 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380066 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380124 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380087 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380068 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380035 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380104 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380082 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380099 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380046 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380034 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06410195 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06502432 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380107 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380108 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380126 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06502429 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380091 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380101 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06454867 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380086 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380070 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380027 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06468854 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380029 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380044 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380112 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380067 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380121 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380117 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06262921 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06262924 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06454856 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06324121 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380122 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380088 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380043 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380123 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380119 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380093 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380078 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380092 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25380110 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10566163 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10566164 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10566167 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10566165 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10566162 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10566166 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1455124_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10566161 | - | - |
Mouse Genome 430 2.0 Array | 1455124_at | - | - |
Mouse Genome 430 2.0 Array | 1455124_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 101700_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 101700_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0700032652.mm.1 | NM_198012.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700032652.mm.1 | TC0700003865.mm.1 | MTA TC |
Mouse Expression Set 430 | 1455124_at | - | - |
Mouse Expression Set 430 | 1455124_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1455124_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1455124_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 874516 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 6940739 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6940739 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006940739 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2635642 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2635642 | - | - |
mouse-6 v1.1 (Illumina) | scl30936.6_1-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6940739 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl30936.6_1-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6940739 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2635642 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2635642 | - | - |
