Assay Details
Gene Symbol: Trim68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC034249.1 | - | 5 - 6 | 959 | 3675490 | 88 |
CO424835.1 | - | 3 - 4 | 438 | 88 | ||
CX220787.1 | - | 2 - 3 | 62 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
