Assay Details
| Gene Symbol: Ctnnd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008729.2 | NP_032755.2 | 14 - 15 | 2986 | 98 | |
| GenBank mRNA | AK140454.1 | - | 4 - 5 | 604 | 98 | |
| BC138260.1 | - | 14 - 15 | 2531 | 8861279 | 98 | |
| BC138261.1 | - | 14 - 15 | 2531 | 8861280 | 98 | |
| BC145217.1 | - | 14 - 15 | 2528 | 9053768 | 98 | |
| U90331.1 | - | 14 - 15 | 2989 | 98 | ||
More Information
Set Membership: |
|

Ctnnd2
Genomic Map