Assay Details
Gene Symbol: Rab19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011226.1 | NP_035356.1 | 1 - 2 | 84 | 99 | |
GenBank mRNA | AK017590.1 | - | 1 - 2 | 118 | 99 | |
BC032936.1 | - | 1 - 2 | 84 | 1379043 | 99 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC032936.1, NM_011226.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
