Assay Details
Gene Symbol: Mars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003913.2 | NP_001003913.1 | 18 - 19 | 2554 | 121 | |
NM_001171582.1 | NP_001165053.1 | 18 - 19 | 2554 | 121 | ||
GenBank mRNA | AA510607.1 | - | 1 - 2 | 120 | 120 | |
BC052042.1 | - | 18 - 19 | 2406 | 121 | ||
BC058968.1 | - | 18 - 19 | 2446 | 121 | ||
BC079643.1 | - | 18 - 19 | 2407 | 6416029 | 121 | |
CJ278192.1 | - | 3 - 4 | 343 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
