Assay Details
Gene Symbol: Mars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003913.2 | NP_001003913.1 | 9 - 10 | 1254 | 65 | |
NM_001171582.1 | NP_001165053.1 | 9 - 10 | 1254 | 65 | ||
GenBank mRNA | BC052042.1 | - | 9 - 10 | 1106 | 65 | |
BC058968.1 | - | 9 - 10 | 1146 | 65 | ||
BC079643.1 | - | 9 - 10 | 1107 | 6416029 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
