Assay Details
| Gene Symbol: Dusp22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134068.3 | NP_598829.1 | 7 - 8 | 579 | 72 | |
| GenBank mRNA | AF237619.1 | - | 7 - 8 | 539 | 72 | |
| CX236611.1 | - | 7 - 8 | 579 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dusp22
Genomic Map