Assay Details
Gene Symbol: Ager | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007425.3 | NP_031451.2 | 8 - 9 | 998 | 58 | |
NR_073173.1 | 8 - 9 | 998 | 58 | |||
NR_073175.1 | 7 - 8 | 1193 | 58 | |||
GenBank mRNA | AB207883.1 | - | 8 - 9 | 961 | 58 | |
BC061182.1 | - | 8 - 9 | 986 | 30300427 | 58 | |
EU520325.1 | - | 8 - 9 | 954 | 58 | ||
EU570240.1 | - | 8 - 9 | 954 | 58 | ||
EU570241.1 | - | 7 - 8 | 1149 | 58 | ||
EU570242.1 | - | 8 - 9 | 954 | 58 | ||
EU570244.1 | - | 7 - 8 | 1149 | 58 | ||
EU570246.1 | - | 7 - 8 | 1111 | 58 | ||
EU906857.1 | - | 3 - 4 | 194 | 58 | ||
EU906864.1 | - | 7 | 1149 | 79 | ||
GU164735.1 | - | 3 - 4 | 213 | 58 | ||
KC692918.1 | - | 8 - 9 | 954 | 58 | ||
L33412.1 | - | 8 - 9 | 962 | 58 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
