Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700006234.mm.1 |
NM_001163379.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006234.mm.1 |
NM_001163379.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006234.mm.1 |
TC1700000639.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700006238.mm.1 |
NM_001163379.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006238.mm.1 |
NM_001163379.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006238.mm.1 |
NM_018862.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006238.mm.1 |
NM_018862.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006238.mm.1 |
TC1700000639.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700006233.mm.1 |
NM_001163379.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006233.mm.1 |
NM_001163379.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006233.mm.1 |
TC1700000639.mm.1 |
MTA TC |