Assay Details
| Gene Symbol: Scube2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020052.2 | NP_064436.2 | 19 - 20 | 2621 | 66 | |
| GenBank mRNA | AK016571.1 | - | 14 - 15 | 1935 | 66 | |
| AK142950.1 | - | 19 - 20 | 2588 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Scube2
Genomic Map