Assay Details
Gene Symbol: Hist1h2ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175660.3 | NP_783591.2 | 1 | 456 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28883287 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28883291 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P420466 | NM_175660.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P420466 | NM_175660.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P420466 | NM_175660.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P420466 | NM_175660.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28883273 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28883276 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28883266 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28883267 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28883274 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28883272 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28883286 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28883291 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28883280 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28883264 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28883261 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28883284 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28883281 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28883287 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28883278 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28883270 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28883279 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28883269 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28883263 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28883285 | - | - |
Whole Mouse Genome G4122A | A_52_P420466 | NM_175660.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P420466 | NM_175660.2 | mouse_transcriptome |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10703447 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10703481 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10703494 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10703477 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10703472 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28883287 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28883291 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883351 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883229 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883220 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883327 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883285 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883216 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883358 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883354 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883273 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883269 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883349 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883215 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883217 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883234 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883356 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883268 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883270 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883348 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883256 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883360 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883248 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883319 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883228 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883264 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883261 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883365 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883353 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883323 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883286 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883325 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883362 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883357 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883280 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883352 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883214 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883218 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883236 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883225 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883368 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883345 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883287 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883274 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883276 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883284 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883330 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883222 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883263 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883332 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883266 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883251 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883252 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883321 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883232 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883281 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28883278 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11114053 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28883273 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28883276 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28883291 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28883270 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28883280 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28883269 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28883279 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28883263 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28883284 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28883264 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28883266 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28883274 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28883272 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28883286 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28883287 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28883278 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28883285 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28883261 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28883281 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10404064 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10404063 | - | - |
Affymetrix Mouse Clariom S Array | PSR1300001453.mm.1 | NM_175660.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300001453.mm.1 | TC1300000260.mm.1 | MTA TC |
Mouse Expression Set 430 | 1418366_PM_at | - | - |
Mouse Expression Set 430 | 1438009_at | - | - |
Mouse Expression Set 430 | 1418367_PM_x_at | - | - |
Mouse Expression Set 430 | 1418366_at | - | - |
Mouse Expression Set 430 | 1438009_PM_at | - | - |
Mouse Expression Set 430 | 1418367_x_at | - | - |
Murine Genome U74v2 Set | 94805_f_at | - | - |
Murine Genome U74v2 Set | 94805_PM_f_at | - | - |
Murine Genome U74v2 Set | 167053_PM_at | - | - |
Murine Genome U74v2 Set | 167053_at | - | - |
Mouse Genome 430A 2.0 Array | 1418366_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1418367_PM_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1418366_at | - | - |
Mouse Genome 430A 2.0 Array | 1418367_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1438009_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1438009_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 897729 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 103120239 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2680465 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 103120239 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002680465 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2704873 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2704873 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1242304 | - | - |
mouse-6 v1.1 (Illumina) | scl12329.1.1_313-S | - | - |
mouse-6 v1.1 (Illumina) | GI_30061368-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2680465 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 103120239 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl12329.1.1_313-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2680465 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103120239 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2704873 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2704873 | - | - |
