Assay Details
Gene Symbol: Lrguk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028886.1 | NP_083162.1 | 11 - 12 | 1345 | 87 | |
GenBank mRNA | AK014976.1 | - | 11 - 12 | 1349 | 87 | |
AK029666.1 | - | 11 - 12 | 1344 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
