Assay Details
Gene Symbol: Lrguk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028886.1 | NP_083162.1 | 1 - 2 | 308 | 59 | |
GenBank mRNA | AK014976.1 | - | 1 - 2 | 312 | 59 | |
AK029666.1 | - | 1 - 2 | 307 | 59 | ||
AK045903.1 | - | 1 - 2 | 308 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P270005 | AK014976.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P270005 | NM_028886.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P270005 | AK014976.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P270005 | NM_028886.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P109080 | AK045903.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 699319 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 285622 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 267138 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 39996 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 552389 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 570763 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 160374 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 480884 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 729145 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 603886 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 765475 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 351335 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 269127 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 542221 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 635537 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 564152 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 467789 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 855519 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 962091 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 419782 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 12335 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 244378 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 426475 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 498741 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 633313 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 906809 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 743740 | AK084548.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 743740 | AK170595.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 743740 | AK172181.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 743740 | BC056486.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 743740 | NM_177394.2 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 190240 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 161952 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 381028 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 534792 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 807511 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 156328 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 748578 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 746204 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 119247 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 658267 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 214897 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 271505 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 150494 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 473032 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 225323 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 639527 | AK045500.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 639527 | AK051821.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 639527 | AK082388.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 639527 | BC052062.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 639527 | NM_172813.2 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 865717 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 165883 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 208395 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 396968 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 7776 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 645025 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 166309 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 723601 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 14214 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 466846 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 444939 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 473675 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 546433 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 748533 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 510477 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 700620 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 375386 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 150850 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 758376 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 227017 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 99857 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 107279 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 95419 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 40487 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 11935 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 125579 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 97649 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 129827 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 137420 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 13093 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 153793 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 18980 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 177082 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 32101 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P109142 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P112525 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P270005 | AK014976.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P270005 | NM_028886.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P270005 | AK014976.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P270005 | NM_028886.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P109080 | AK045903.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05858610 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05858657 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05858625 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05858531 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P112525 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05858408 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05858504 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P109142 | - | - |
Whole Mouse Genome G4122A | A_51_P270005 | AK014976.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P270005 | NM_028886.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P270005 | AK014976.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P270005 | NM_028886.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P112525 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05858657 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P109142 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05858504 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05370552 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353209 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353505 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05863957 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353296 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353262 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353517 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353248 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353175 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353413 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353274 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353411 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353393 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353371 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353384 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353375 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353932 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353395 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353180 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353409 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353929 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353922 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353278 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353264 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353211 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353223 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353257 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05864235 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353164 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353928 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353434 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353189 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05861676 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353156 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353416 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353389 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05822326 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05810553 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05743056 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353261 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353444 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353195 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353506 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353151 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353398 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353381 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353206 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05177672 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353443 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05705790 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353448 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353229 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353516 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05547654 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353152 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353162 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353383 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05098316 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353912 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353253 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353336 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353170 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353280 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353507 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353272 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353519 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353283 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05037884 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353510 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353191 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05185821 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353514 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353388 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05036801 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05827783 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353437 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353165 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353193 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353364 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353345 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353213 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353377 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353157 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05089047 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353231 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353919 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353207 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353247 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353419 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353368 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353155 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353382 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353210 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05863952 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05036800 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353166 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353930 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353288 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05571980 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05571978 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353230 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353275 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05592467 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353235 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353224 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353182 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353201 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353149 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353217 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353339 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353221 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353290 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353363 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05751023 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353297 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05058971 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353226 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05009080 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05690634 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353352 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353436 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353295 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353343 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353348 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353340 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353240 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353276 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353370 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353167 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353158 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353376 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353442 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05751022 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353911 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353445 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353350 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353285 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353446 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05836338 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353161 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353176 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353220 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353916 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05861681 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353908 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353203 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353418 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353178 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353391 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353244 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05743053 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353259 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353439 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353396 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353513 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05367222 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353362 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353179 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353420 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353917 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353184 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353405 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353159 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P24353233 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1430217_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10537123 | - | - |
Mouse Genome 430 2.0 Array | 1430217_at | - | - |
Mouse Genome 430 2.0 Array | 1430217_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74354_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74354_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0600002326.mm.1 | TC0600000319.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600002326.mm.1 | NM_028886.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600002326.mm.1 | NM_028886.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600002325.mm.1 | TC0600000319.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0600002325.mm.1 | NM_028886.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0600002325.mm.1 | NM_028886.1 | Refseq |
Mouse Expression Set 430 | 1430217_PM_at | - | - |
Mouse Expression Set 430 | 1430217_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1430217_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1430217_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 448152 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 105860176 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 105860176 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2693661 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2693661 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1240483 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 105860176 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105860176 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1240483 | - | - |
