Assay Details
Gene Symbol: Lrguk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028886.1 | NP_083162.1 | 3 - 4 | 499 | 123 | |
GenBank mRNA | AK014976.1 | - | 3 - 4 | 503 | 123 | |
AK029666.1 | - | 3 - 4 | 498 | 123 | ||
AK045903.1 | - | 3 - 4 | 499 | 123 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
