Assay Details
| Gene Symbol: Dnajc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009584.4 | NP_033610.1 | 13 - 14 | 1536 | 140 | |
| GenBank mRNA | AK131964.1 | - | 13 - 14 | 1557 | 140 | |
| BC021755.1 | - | 12 - 13 | 3281 | 140 | ||
| BC052027.1 | - | 13 - 14 | 1563 | 5718637 | 140 | |
| D63784.1 | - | 13 - 14 | 1535 | 140 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dnajc2
Genomic Map