Assay Details
| Gene Symbol: Dnajc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009584.4 | NP_033610.1 | 3 - 4 | 445 | 70 | |
| GenBank mRNA | AK013295.1 | - | 3 - 4 | 436 | 70 | |
| AK131964.1 | - | 3 - 4 | 466 | 70 | ||
| AK136951.1 | - | 3 - 4 | 469 | 70 | ||
| AK161347.1 | - | 3 - 4 | 412 | 70 | ||
| AK161376.1 | - | 3 - 4 | 466 | 70 | ||
| AK162409.1 | - | 3 - 4 | 466 | 70 | ||
| AK167991.1 | - | 3 - 4 | 466 | 70 | ||
| AK169850.1 | - | 3 - 4 | 480 | 70 | ||
| BC021755.1 | - | 3 - 4 | 450 | 70 | ||
| BC052027.1 | - | 3 - 4 | 472 | 5718637 | 70 | |
| D63784.1 | - | 3 - 4 | 444 | 70 | ||
| U53208.1 | - | 3 - 4 | 400 | 70 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dnajc2
Genomic Map