Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600001160.mm.1 |
TC0600000182.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600001160.mm.1 |
NM_001081351.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600001160.mm.1 |
NM_001081351.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600001161.mm.1 |
TC0600000182.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600001161.mm.1 |
NM_001081351.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600001161.mm.1 |
NM_001081351.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600001170.mm.1 |
TC0600000182.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600001170.mm.1 |
NM_001081351.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600001170.mm.1 |
NM_001081351.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600001162.mm.1 |
TC0600000182.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600001162.mm.1 |
NM_001081351.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600001162.mm.1 |
NM_001081351.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600001165.mm.1 |
TC0600000182.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600001165.mm.1 |
NM_001081351.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600001165.mm.1 |
NM_001081351.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600001163.mm.1 |
TC0600000182.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600001163.mm.1 |
NM_001081351.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600001163.mm.1 |
NM_001081351.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600001164.mm.1 |
TC0600000182.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600001164.mm.1 |
NM_001081351.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600001164.mm.1 |
NM_001081351.1 |
Refseq |