Assay Details
Gene Symbol: Lnpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172827.3 | NP_766415.1 | 14 - 15 | 2635 | 69 | |
GenBank mRNA | AK029094.1 | - | 14 - 15 | 2636 | 69 | |
AK161484.1 | - | 1862 | 69 | |||
BC120925.1 | - | 14 - 15 | 2592 | 40098425 | 69 | |
BC120926.1 | - | 14 - 15 | 2592 | 40098428 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
