Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700013545.mm.1 |
NM_172827.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013545.mm.1 |
NM_172827.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013545.mm.1 |
TC1700001516.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700013550.mm.1 |
NM_172827.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013550.mm.1 |
NM_172827.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013550.mm.1 |
TC1700001516.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700013551.mm.1 |
NM_172827.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013551.mm.1 |
NM_172827.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013551.mm.1 |
TC1700001516.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700013546.mm.1 |
NM_172827.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013546.mm.1 |
NM_172827.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700013546.mm.1 |
TC1700001516.mm.1 |
MTA TC |