Assay Details
| Gene Symbol: Lrp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172814.3 | NP_766402.1 | 3 - 4 | 337 | 76 | |
| GenBank mRNA | AK050512.1 | - | 3 - 4 | 338 | 76 | |
| BC058345.1 | - | 2 - 3 | 517 | 6822657 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrp12
Genomic Map