Assay Details
| Gene Symbol: Stk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019635.2 | NP_062609.2 | 3 - 4 | 485 | 131 | |
| GenBank mRNA | AF271361.1 | - | 3 - 4 | 231 | 131 | |
| AY058922.1 | - | 3 - 4 | 371 | 131 | ||
| BC049123.1 | - | 3 - 4 | 359 | 6813956 | 131 | |
| U28726.1 | - | 3 - 4 | 364 | 131 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stk3
Genomic Map