Assay Details
| Gene Symbol: Stk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019635.2 | NP_062609.2 | 9 - 10 | 1408 | 153 | |
| GenBank mRNA | AF271361.1 | - | 9 - 10 | 1154 | 153 | |
| AK002914.1 | - | 4 - 5 | 626 | 153 | ||
| AY058922.1 | - | 9 - 10 | 1294 | 153 | ||
| BC037440.1 | - | 7 - 8 | 1164 | 3585312 | 153 | |
| BC049123.1 | - | 9 - 10 | 1282 | 6813956 | 153 | |
| U28726.1 | - | 9 - 10 | 1287 | 153 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stk3
Genomic Map