Assay Details
Gene Symbol: Xylt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175645.3 | NP_783576.2 | 8 - 9 | 1949 | 72 | |
GenBank mRNA | AJ291750.1 | - | 6 - 7 | 1266 | 72 | |
AJ539164.1 | - | 8 - 9 | 1951 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
